Assay Details
Target Gene Details
Entrez Gene ID: | 5148 |
Gene Name: | phosphodiesterase 6G |
Gene Aliases: |
PDEG, RP57 |
Location: |
Chr.17:81650459-81663418 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE6G | NM_002602.3 | NP_002593.1 | ||
NR_026872.1 | ||||
XM_006721942.3 | XP_006722005.1 | |||
XM_017024734.1 | XP_016880223.1 | |||
XM_017024735.1 | XP_016880224.1 | |||
XM_017024736.1 | XP_016880225.1 | |||
BQ639341.1 | ||||
BX537414.1 | CAD97656.1 | |||
BX647779.1 | ||||
M36476.1 | AAA60044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | ARL16 LOC100129503 TSPAN10 OXLD1 MIR3186 LOC107985021 MIR6786 LINC00482 ACTG1 CHMP6 FSCN2 MRPL12 AATK-AS1 MIR4740 TEPSIN HGS MIR338 BAIAP2-AS1 BAHCC1 SLC25A10 BAIAP2 AATK NPLOC4 C17orf89 LOC105371921 CEP131 MIR657 CCDC137 MIR3065 MIR1250 SLC38A10 LOC100130370 PDE6G LOC107985022 TMEM105 LOC101928855 LOC400627 RPTOR LOC105371925 FAAP100 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | ARL16 ALYREF DUS1L NOTUM OXLD1 C17orf62 ASPSCR1 MIR6786 LINC00482 NPB FSCN2 MRPL12 AATK-AS1 MAFG GPS1 FN3KRP HGS ARHGDIA CSNK1D C17orf89 PCYT2 MAFG-AS1 CCDC137 SECTM1 MCRIP1 RAB40B CD7 ZNF750 LOC105376790 PDE6G LOC105376791 FASN ANAPC11 TEX19 HEXDC FAAP100 STRA13 RAC3 PYCR1 TSPAN10 MIR3186 SNORD134 ACTG1 B3GNTL1 WDR45B LOC101929552 MIR4740 TEPSIN LRRC45 SLC16A3 BAHCC1 DCXR SLC25A10 LOC105371944 RFNG AATK NPLOC4 GCGR TBCD CCDC57 FN3K P4HB OGFOD3 MYADML2 CEP131 SIRT7 LOC101929511 UTS2R NARF SLC38A10 PPP1R27 FOXK2 LOC100130370 TMEM105 MIR6787 LOC105371925 MIR4525 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | STRA13 ARL16 ALYREF RAC3 PYCR1 DUS1L NOTUM TSPAN10 OXLD1 ASPSCR1 MIR6786 NPB FSCN2 MRPL12 MAFG GPS1 HGS LRRC45 ARHGDIA DCXR SLC25A10 RFNG NPLOC4 GCGR PCYT2 P4HB MYADML2 SIRT7 MAFG-AS1 CCDC137 MCRIP1 PPP1R27 PDE6G ANAPC11 FAAP100 |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | ARL16 CCDC137 TSPAN10 OXLD1 MIR6786 MCRIP1 PPP1R27 MRPL12 PDE6G HGS SLC25A10 GCGR P4HB |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |