Assay Details
Target Gene Details
Entrez Gene ID: | 1089 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 4 |
Gene Aliases: |
CGM7, CGM7_HUMAN, NCA |
Location: |
Chr.19:41616986-41627087 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2718577 | Chr.19:41576456 - 41709018 on Build GRCh38 | Deletion | LOC105372404 CEACAM5 CEACAM21 CEACAM7 CEACAM4 |
nsv519408 | Chr.19:41559909 - 41667978 on Build GRCh38 | Gain | LOC105372404 CEACAM21 CEACAM4 |
nsv2494 | Chr.19:41612811 - 41648542 on Build GRCh38 | Insertion | CEACAM4 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | LOC101930071 POU2F2 MEGF8 MIR4323 MIR6797 CD79A GSK3A RPS19 ARHGEF1 MIR8077 CIC CNFN LOC390937 CEACAM3 DMRTC2 CEACAM6 LOC100505585 TMEM145 GRIK5 LOC100505622 ATP1A3 CEACAM4 PAFAH1B3 CEACAM5 PRR19 LIPE-AS1 LYPD4 RABAC1 CEACAM7 ZNF574 ERF ZNF526 DEDD2 LIPE |
dgv3549n100 | Chr.19:41550966 - 41671232 on Build GRCh38 | Gain | LOC105372404 CEACAM21 CEACAM4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |