Assay Details
Target Gene Details
Entrez Gene ID: | 341567 |
Gene Name: | H1 histone family member N, testis specific |
Gene Aliases: |
H1.7, H1T2 |
Location: |
Chr.12:48328980-48330279 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H1FNT | NM_181788.1 | 1 | 1006 | NP_861453.1 |
AB121028.2 | 1 | 646 | ||
AY302593.1 | 1 | 1006 | AAP60022.1 | |
BC118635.1 | 1 | 954 | ||
BC119787.1 | 1 | 956 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss | CCDC184 ASB8 ZNF641 PFKM MIR6505 SENP1 OR10AD1 H1FNT C12orf54 |
nsv1160031 | Chr.12:48277081 - 48330531 on Build GRCh38 | Deletion | H1FNT |
nsv437752 | Chr.12:48328049 - 48347436 on Build GRCh38 | Loss | ZNF641 H1FNT |
nsv832402 | Chr.12:48292978 - 48457237 on Build GRCh38 | Gain | ZNF641 H1FNT C12orf54 |
esv3629436 | Chr.12:48320207 - 48364691 on Build GRCh38 | Gain | ZNF641 H1FNT |
nsv826362 | Chr.12:48299265 - 48375070 on Build GRCh38 | Gain | ZNF641 H1FNT |
nsv8971 | Chr.12:48142743 - 48364718 on Build GRCh38 | Gain+Loss | CCDC184 ASB8 ZNF641 PFKM OR10AD1 H1FNT |
esv34439 | Chr.12:48147974 - 48385779 on Build GRCh38 | Gain | CCDC184 ASB8 ZNF641 OR10AD1 H1FNT |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |