Assay Details
Target Gene Details
Entrez Gene ID: | 23582 |
Gene Name: | cyclin D1 binding protein 1 |
Gene Aliases: |
DIP1, GCIP, HHM |
Location: |
Chr.15:43185118-43197177 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCNDBP1 | NM_012142.4 | 5 | 613 | NP_036274.3 |
NR_027513.2 | ||||
NR_027514.2 | ||||
NR_045998.1 | 4 | 533 | ||
NR_045999.1 | ||||
XM_006720448.1 | 3 | 535 | XP_006720511.1 | |
AF082569.1 | 5 | 472 | AAD11777.1 | |
AF087852.1 | 5 | 458 | AAP97163.1 | |
AF113535.1 | 3 | 422 | AAF14872.1 | |
AF132034.1 | 5 | 428 | AAF77613.1 | |
AF246144.1 | 5 | 430 | AAF67182.1 | |
AK075146.1 | 5 | 442 | BAC11433.1 | |
AK075296.1 | 3 | 427 | BAC11530.1 | |
AK124193.1 | 1 | 844 | ||
AK128849.1 | 1 | 1654 | BAC87645.1 | |
AK225143.1 | 5 | 428 | ||
AK225645.1 | 5 | 442 | ||
AK290665.1 | 5 | 442 | ||
AK290707.1 | 4 | 395 | ||
AK295017.1 | ||||
AK308817.1 | 4 | 1037 | ||
AL552334.3 | 5 | 552 | ||
BC009689.1 | 5 | 456 | AAH09689.1 | |
BQ439439.1 | ||||
CR450331.1 | 5 | 382 | CAG29327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569241 | Chr.15:42908911 - 43230724 on Build GRCh38 | Loss | UBR1 CCNDBP1 TMEM62 EPB42 TTBK2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |