Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | ACTG1 MIR338 MIR3065 BAHCC1 LOC100129503 SLC25A10 AATK NPLOC4 HGS CEP131 TSPAN10 MIR6786 C17orf89 ARL16 BAIAP2 BAIAP2-AS1 TEPSIN CHMP6 OXLD1 LOC100130370 LOC107985021 MIR4740 FSCN2 LOC400627 MIR3186 LOC105371925 LOC105371921 RPTOR LOC101928855 MIR1250 MRPL12 FAAP100 MIR657 SLC38A10 AATK-AS1 TMEM105 LOC107985022 LINC00482 PDE6G CCDC137 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | CEP131 AATK-AS1 AATK |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | PYCR1 MIR6787 GCGR ZNF750 FN3KRP SLC16A3 AATK NPLOC4 HGS STRA13 TSPAN10 LOC101929511 MIR6786 NOTUM RAB40B LOC105376791 RFNG OXLD1 CCDC57 DUS1L SECTM1 LOC101929552 MIR3186 HEXDC LOC105376790 ALYREF MRPL12 FAAP100 NARF MAFG UTS2R GPS1 SLC38A10 TBCD PDE6G MCRIP1 FN3K MYADML2 CCDC137 CD7 SNORD134 LOC105371944 ACTG1 LRRC45 TEX19 BAHCC1 SLC25A10 FASN CEP131 C17orf89 ARL16 CSNK1D ARHGDIA TEPSIN WDR45B LOC100130370 SIRT7 MIR4740 FSCN2 B3GNTL1 P4HB LOC105371925 RAC3 PCYT2 FOXK2 ASPSCR1 MIR4525 PPP1R27 NPB DCXR OGFOD3 AATK-AS1 TMEM105 MAFG-AS1 LINC00482 C17orf62 ANAPC11 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | CEP131 SLC38A10 AATK-AS1 C17orf89 LOC105371925 TEPSIN AATK |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | CEP131 MIR657 AATK-AS1 MIR338 MIR3065 BAIAP2 AATK MIR1250 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | MIR338 LOC105371925 MIR3065 AATK MIR1250 CEP131 MIR657 SLC38A10 AATK-AS1 TMEM105 C17orf89 LINC00482 BAIAP2 BAIAP2-AS1 TEPSIN |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | CEP131 MIR657 AATK-AS1 MIR338 MIR3065 BAIAP2 BAIAP2-AS1 AATK MIR1250 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | LOC100130370 MIR4740 FSCN2 ACTG1 MIR3186 LOC105371925 BAHCC1 AATK NPLOC4 FAAP100 CEP131 SLC38A10 AATK-AS1 TMEM105 C17orf89 LINC00482 TEPSIN |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR657 AATK-AS1 MIR338 MIR3065 BAIAP2 AATK MIR1250 |