Assay Details
Target Gene Details
Entrez Gene ID: | 79974 |
Gene Name: | cadherin like and PC-esterase domain containing 1 |
Gene Aliases: |
C7orf58 |
Location: |
Chr.7:120988697-121297444 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPED1 | NM_001105533.1 | 1 | 397 | NP_001099003.1 |
NM_024913.4 | 2 | 613 | NP_079189.4 | |
XM_017012649.1 | 1 | 1089 | XP_016868138.1 | |
AK055624.1 | 1 | 737 | BAB70974.1 | |
AK289978.1 | 2 | 610 | ||
AL832619.2 | 2 | 568 | CAD89961.1 | |
AL832638.2 | 2 | 437 | CAD89953.1 | |
AY358220.1 | 1 | 189 | AAQ88587.1 | |
AY780785.1 | 1 | 166 | AAV98355.1 | |
BC046360.1 | 1 | 613 | AAH46360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831114 | Chr.7:120955570 - 121164456 on Build GRCh38 | Loss | CPED1 ING3 |