Assay Details
Target Gene Details
Entrez Gene ID: | 83450 |
Gene Name: | dynein regulatory complex subunit 3 |
Gene Aliases: |
CFAP134, LRRC48 |
Location: |
Chr.17:17972766-18016889 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DRC3 | NM_001130090.1 | 4 | 418 | NP_001123562.1 |
NM_001130091.1 | 4 | 418 | NP_001123563.1 | |
NM_001130092.1 | 3 | 329 | NP_001123564.1 | |
NM_031294.3 | 3 | 329 | NP_112584.3 | |
XM_006721583.3 | 1 | 56 | XP_006721646.1 | |
XM_011524018.2 | 3 | 361 | XP_011522320.1 | |
XM_011524019.2 | 3 | 207 | XP_011522321.1 | |
XM_011524020.2 | 3 | 486 | XP_011522322.1 | |
XM_011524021.2 | 3 | 359 | XP_011522323.1 | |
XM_011524022.2 | 3 | 368 | XP_011522324.1 | |
XM_011524023.2 | 3 | 368 | XP_011522325.1 | |
AK093317.1 | 2 | 180 | ||
AK293011.1 | 4 | 297 | ||
AL136926.1 | 3 | 240 | CAB66860.2 | |
BC040276.1 | 3 | 205 | AAH40276.1 | |
BC047873.1 | 3 | 257 | AAH47873.1 | |
BC050419.1 | 4 | 279 | AAH50419.1 | |
BC050665.1 | 4 | 275 | AAH50665.1 | |
DB032678.1 | 3 | 329 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510701 | Chr.17:17947160 - 18052171 on Build GRCh38 | Deletion | DRC3 ATPAF2 TOM1L2 GID4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |