Assay Details
Target Gene Details
Entrez Gene ID: | 100529251 |
Gene Name: | CKLF-CMTM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:66552563-66579135 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CKLF-CMTM1 | NM_001202509.1 | NP_001189438.1 | ||
NM_001204098.1 | NP_001191027.1 | |||
NM_001204099.1 | NP_001191028.1 | |||
BM929895.1 | ||||
FY211334.1 | ||||
FY212695.1 |
Target Gene Details
Entrez Gene ID: | 113540 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 1 |
Gene Aliases: |
CKLFH, CKLFH1, CKLFSF1 |
Location: |
Chr.16:66566391-66579135 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM1 | NM_052999.3 | 1 | 83 | NP_443725.3 |
NM_181268.2 | 1 | 83 | NP_851785.2 | |
NM_181269.2 | 1 | 139 | NP_851786.1 | |
NM_181270.2 | 1 | 139 | NP_851787.1 | |
NM_181271.2 | 1 | 139 | NP_851788.1 | |
NM_181272.2 | 1 | 139 | NP_851789.1 | |
NM_181283.2 | 1 | 139 | NP_851800.1 | |
NM_181296.2 | 1 | 139 | NP_851813.1 | |
AY174118.1 | 1 | 140 | AAN73227.1 | |
AY174119.1 | 1 | 140 | AAN73228.1 | |
AY174120.1 | 1 | 140 | AAN73229.1 | |
AY174121.1 | 1 | 140 | AAN73230.1 | |
AY174122.1 | 1 | 140 | AAN73231.1 | |
AY174123.1 | 1 | 140 | AAN73232.1 | |
AY174124.1 | 1 | 140 | AAN73233.1 | |
AY174125.1 | 1 | 140 | AAN73234.1 | |
AY174126.1 | 1 | 140 | AAN73235.1 | |
AY174127.1 | 1 | 140 | AAN73236.1 | |
AY174128.1 | 1 | 140 | AAN73237.1 | |
AY174129.1 | AAN73238.1 | |||
AY174130.1 | 1 | 140 | AAN73239.1 | |
AY174131.1 | 1 | 140 | AAN73240.1 | |
AY174132.1 | 1 | 140 | AAN73241.1 | |
AY174133.1 | 1 | 140 | AAN73242.1 | |
BC057852.1 | 1 | 83 | AAH57852.2 | |
DN992496.1 | 1 | 285 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |