Assay Details
Target Gene Details
Entrez Gene ID: | 201633 |
Gene Name: | T-cell immunoreceptor with Ig and ITIM domains |
Gene Aliases: |
VSIG9, VSTM3, WUCAM |
Location: |
Chr.3:114293986-114310288 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIGIT | NM_173799.3 | 4 | 768 | NP_776160.2 |
XM_011512538.1 | 4 | 753 | XP_011510840.1 | |
XM_017005865.1 | 3 | 1043 | XP_016861354.1 | |
AK097192.1 | 4 | 726 | BAC04973.1 | |
AL833175.1 | 5 | 3508 | CAI46183.1 | |
BC101288.1 | 4 | 725 | AAI01289.1 | |
BC101289.1 | 4 | 725 | AAI01290.1 | |
BC101290.1 | 4 | 725 | AAI01291.1 | |
BC101291.1 | 4 | 725 | AAI01292.1 | |
BX640915.1 | 3 | 647 | CAE45956.1 | |
EU675310.1 | 4 | 768 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |