Assay Details
Target Gene Details
Entrez Gene ID: | 572 |
Gene Name: | BCL2 associated agonist of cell death |
Gene Aliases: |
BBC2, BCL2L8 |
Location: |
Chr.11:64269828-64284704 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAD | NM_004322.3 | NP_004313.1 | ||
NM_032989.2 | NP_116784.1 | |||
AF021792.1 | AAB72092.1 | |||
AK291863.1 | ||||
AK303051.1 | ||||
AW014069.1 | ||||
BC001901.2 | AAH01901.1 | |||
BC095431.1 | AAH95431.1 | |||
U66879.1 | AAB36516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | GPR137 PLCB3 DNAJC4 STIP1 BAD NUDT22 MACROD1 LOC105369340 FKBP2 PPP1R14B TRPT1 FERMT3 VEGFB |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | GPR137 PRDX5 PLCB3 STIP1 TEX40 LOC105369340 ESRRA KCNK4-TEX40 FERMT3 OTUB1 FLRT1 MIR7155 DNAJC4 BAD NUDT22 TRMT112 COX8A MACROD1 FKBP2 PPP1R14B TRPT1 KCNK4 NAA40 VEGFB CCDC88B |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | GPR137 PRDX5 PLCB3 DNAJC4 STIP1 BAD NUDT22 TRMT112 TEX40 LOC105369340 FKBP2 ESRRA PPP1R14B KCNK4-TEX40 TRPT1 FERMT3 KCNK4 VEGFB |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | GPR137 PRDX5 PLCB3 DNAJC4 BAD NUDT22 TRMT112 TEX40 LOC105369340 FKBP2 RPS6KA4 ESRRA MIR1237 PPP1R14B KCNK4-TEX40 TRPT1 FERMT3 KCNK4 VEGFB MIR7155 CCDC88B |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | GPR137 PLCB3 DNAJC4 STIP1 BAD NUDT22 MACROD1 LOC105369340 FKBP2 PPP1R14B TRPT1 FERMT3 VEGFB |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | GPR137 PRDX5 PLCB3 DNAJC4 STIP1 BAD NUDT22 TRMT112 TEX40 MACROD1 LOC105369340 FKBP2 ESRRA PPP1R14B KCNK4-TEX40 TRPT1 FERMT3 KCNK4 FLRT1 VEGFB |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |