Assay Details
Target Gene Details
Entrez Gene ID: | 2829 |
Gene Name: | X-C motif chemokine receptor 1 |
Gene Aliases: |
CCXCR1, GPR5 |
Location: |
Chr.3:46016990-46086803 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XCR1 | NM_001024644.1 | 2 | 674 | NP_001019815.1 |
NM_005283.2 | 3 | 796 | NP_005274.1 | |
XM_017006196.1 | 4 | 1814 | XP_016861685.1 | |
XM_017006197.1 | 6 | 2066 | XP_016861686.1 | |
XM_017006198.1 | 5 | 1879 | XP_016861687.1 | |
XM_017006199.1 | 4 | 876 | XP_016861688.1 | |
XM_017006200.1 | 3 | 1734 | XP_016861689.1 | |
XM_017006201.1 | 2 | 1583 | XP_016861690.1 | |
AK314277.1 | 2 | 927 | ||
BC069075.1 | 1 | 572 | AAH69075.1 | |
BC069589.1 | 1 | 572 | AAH69589.1 | |
BC095533.1 | 1 | 623 | AAH95533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759145 | Chr.3:45974972 - 46078908 on Build GRCh38 | Loss | XCR1 FYCO1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |