Assay Details
Target Gene Details
Entrez Gene ID: | 1185 |
Gene Name: | chloride voltage-gated channel 6 |
Gene Aliases: |
CLC-6 |
Location: |
Chr.1:11806096-11843144 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLCN6 | NM_001256959.1 | 22 | 3101 | NP_001243888.1 |
NM_001286.3 | 23 | 3167 | NP_001277.1 | |
NR_046428.1 | 23 | 3151 | ||
AK289999.1 | 23 | 3072 | ||
AK294764.1 | 22 | 2956 | ||
D28475.2 | 23 | 3052 | ||
X83378.1 | 23 | 3026 | CAA58292.1 |
Target Gene Details
Entrez Gene ID: | 100379251 |
Gene Name: | NPPA antisense RNA 1 |
Gene Aliases: |
NPPA-AS, NPPAAS |
Location: |
Chr.1:11840319-11847616 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPPA-AS1 | NR_037806.1 | 1 | 295 | |
FJ706070.1 | 1 | 295 | ||
FJ706071.1 | 1 | 295 | ||
FJ706072.1 | ||||
FJ706073.1 | ||||
FJ706074.1 | ||||
FJ706075.1 | ||||
FJ706076.1 | ||||
FJ706077.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |