Assay Details
Target Gene Details
Entrez Gene ID: | 3586 |
Gene Name: | interleukin 10 |
Gene Aliases: |
CSIF, GVHDS, IL-10, IL10A, TGIF |
Location: |
Chr.1:206767603-206772494 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL10 | NM_000572.2 | 1 | 68 | NP_000563.1 |
AY029171.1 | AAK38162.1 | |||
BC104252.1 | 1 | 65 | AAI04253.1 | |
BC104253.1 | 1 | 65 | AAI04254.1 | |
CR541993.1 | CAG46790.1 | |||
CR542028.1 | CAG46825.1 | |||
EU751618.2 | ||||
EU751619.2 | ||||
M57627.1 | AAA63207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3578391 | Chr.1:206770892 - 206779577 on Build GRCh38 | Loss | IL10 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |