Assay Details
Target Gene Details
Entrez Gene ID: | 57212 |
Gene Name: | TP73 antisense RNA 1 |
Gene Aliases: |
KIAA0495, PDAM |
Location: |
Chr.1:3735984-3747373 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP73-AS1 | NR_033708.1 | 5 | 2685 | |
NR_033709.1 | 5 | 3298 | ||
NR_033710.1 | 5 | 4553 | ||
NR_033711.1 | 4 | 5017 | ||
NR_033712.1 | 6 | 1932 | ||
AB007964.1 | 4 | 4967 | ||
AL133648.1 | 5 | 3806 | CAB63767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460061 | Chr.1:3680453 - 3755227 on Build GRCh38 | Loss | TP73-AS1 CCDC27 TP73 |
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss | MEGF6 TPRG1L PRDM16 TP73-AS1 WRAP73 ARHGEF16 CCDC27 MIR551A TP73 |
esv994567 | Chr.1:3735559 - 3738806 on Build GRCh38 | Deletion | TP73-AS1 TP73 |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss | TPRG1L TP73-AS1 WRAP73 CCDC27 TP73 |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain | SMIM1 MEGF6 TPRG1L LINC01134 TP73-AS1 C1orf174 LINC01346 WRAP73 LRRC47 CCDC27 CEP104 TP73 DFFB |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain | SMIM1 MEGF6 TPRG1L LINC01134 AJAP1 TP73-AS1 MIR551A C1orf174 LOC284661 LINC01346 WRAP73 ARHGEF16 LRRC47 CCDC27 CEP104 TP73 DFFB |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion | SMIM1 MEGF6 LOC105378604 TPRG1L TP73-AS1 MIR4251 MIR551A LINC00982 PRDM16 WRAP73 ARHGEF16 LRRC47 CCDC27 TP73 |
nsv830314 | Chr.1:3660776 - 3804275 on Build GRCh38 | Gain+Loss | SMIM1 TP73-AS1 LRRC47 CCDC27 TP73 |
dgv13n100 | Chr.1:3730448 - 3791316 on Build GRCh38 | Gain | SMIM1 TP73-AS1 LRRC47 CCDC27 TP73 |
esv2758916 | Chr.1:3668477 - 3738972 on Build GRCh38 | Gain | TP73-AS1 TP73 |