Assay Details
Target Gene Details
Entrez Gene ID: | 593 |
Gene Name: | branched chain keto acid dehydrogenase E1, alpha polypeptide |
Gene Aliases: |
BCKDE1A, MSU, MSUD1, OVD1A |
Location: |
Chr.19:41397789-41425005 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCKDHA | NM_000709.3 | NP_000700.1 | ||
NM_001164783.1 | NP_001158255.1 | |||
AB209828.1 | BAD93065.1 | |||
AK298188.1 | ||||
AK313300.1 | ||||
AL833143.1 | CAH56215.1 | |||
BC007878.2 | AAH07878.1 | |||
BC008933.2 | AAH08933.1 | |||
BC023983.1 | AAH23983.1 | |||
BI910860.1 | ||||
J04474.1 | AAB59549.1 | |||
M22221.1 | AAA35590.1 | |||
Z14093.1 | CAA78475.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |