Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | 4 | 368 | NP_001305309.1 |
NM_177533.4 | 4 | 368 | NP_803877.2 | |
AB087802.1 | 4 | 254 | BAC65455.1 | |
AK128196.1 | 4 | 357 | ||
AK310449.1 | 4 | 345 | ||
BC041584.1 | 4 | 330 | AAH41584.1 | |
DA249638.1 | 4 | 288 | ||
DA409636.1 | 4 | 351 | ||
DB096906.1 | 4 | 370 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | MIR6765 NUDT14 JAG2 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | NUDT14 JAG2 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | MIR6765 BRF1 PACS2 LOC102723354 TEX22 BTBD6 NUDT14 MTA1 GPR132 CRIP2 LOC100507437 JAG2 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | C14orf80 PACS2 IGHA2 CRIP1 BTBD6 MTA1 LOC100507437 TMEM121 JAG2 MIR6765 BRF1 IGH TEX22 NUDT14 LOC105370697 CRIP2 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | MIR6765 BRF1 LOC102723354 NUDT14 GPR132 JAG2 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | MIR6765 BRF1 NUDT14 JAG2 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | MIR6765 NUDT14 JAG2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LOC107984648 LOC105370691 LINC00638 XRCC3 CEP170B TRMT61A PPP1R13B AHNAK2 MIR203A GPR132 ADSSL1 JAG2 BAG5 MIR6765 MIR203B LINC00637 BRF1 LOC102723354 ZFYVE21 NUDT14 ASPG CDCA4 SNORA28 C14orf180 LOC388022 SIVA1 MARK3 KIF26A EIF5 PACS2 BTBD6 INF2 CKB LOC101929634 LOC105378183 RD3L MIR4710 TDRD9 ZBTB42 PLD4 C14orf2 KLC1 AKT1 C14orf79 TMEM179 LOC105370708 LOC105370687 APOPT1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | MIR6765 BRF1 LOC102723354 BTBD6 NUDT14 AHNAK2 C14orf79 CDCA4 GPR132 JAG2 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGHV3-71 IGHG1 IGHV3-48 ELK2AP MIR8071-2 IGHG4 IGHV3-30 IGHV3-69-1 LOC100507437 TMEM121 LINC00221 MIR4537 JAG2 MIR6765 BRF1 IGHV1-69 IGH TEX22 NUDT14 IGHV4-55 MIR4538 IGHV3-23 CRIP2 IGHD C14orf80 IGHV4-31 PACS2 IGHA2 CRIP1 BTBD6 MTA1 IGHA1 MIR4507 MIR4539 IGHV3-41 LOC107987211 KIAA0125 LINC00226 IGHG2 LOC105370697 MIR8071-1 IGHG3 ADAM6 IGHM |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | MIR6765 BRF1 LOC102723354 NUDT14 AHNAK2 C14orf79 CDCA4 GPR132 JAG2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |