Assay Details
Target Gene Details
Entrez Gene ID: | 201633 |
Gene Name: | T-cell immunoreceptor with Ig and ITIM domains |
Gene Aliases: |
VSIG9, VSTM3, WUCAM |
Location: |
Chr.3:114293986-114310288 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIGIT | NM_173799.3 | 2 | 223 | NP_776160.2 |
XM_011512538.1 | 2 | 211 | XP_011510840.1 | |
AK097192.1 | 2 | 181 | BAC04973.1 | |
AL833175.1 | 3 | 2963 | CAI46183.1 | |
BC101288.1 | 2 | 180 | AAI01289.1 | |
BC101289.1 | 2 | 180 | AAI01290.1 | |
BC101290.1 | 2 | 180 | AAI01291.1 | |
BC101291.1 | 2 | 180 | AAI01292.1 | |
BX640915.1 | 1 | 87 | CAE45956.1 | |
EU675310.1 | 2 | 223 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv921e214 | Chr.3:114256209 - 114297720 on Build GRCh38 | Gain | TIGIT |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |