Assay Details
Target Gene Details
Entrez Gene ID: | 253558 |
Gene Name: | lysocardiolipin acyltransferase 1 |
Gene Aliases: |
1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849 |
Location: |
Chr.2:30447226-30644225 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCLAT1 | NM_001002257.2 | 1 | 67 | NP_001002257.1 |
NM_001304445.1 | 1 | 67 | NP_001291374.1 | |
NM_001304446.1 | 1 | 67 | NP_001291375.1 | |
NM_182551.4 | 1 | 67 | NP_872357.2 | |
XM_005264244.1 | 1 | 50 | XP_005264301.1 | |
XM_005264245.3 | 1 | 52 | XP_005264302.1 | |
XM_011532744.2 | 1 | 62 | XP_011531046.1 | |
XM_011532745.1 | 1 | 50 | XP_011531047.1 | |
XM_017003746.1 | 1 | 45 | XP_016859235.1 | |
XM_017003747.1 | 1 | 50 | XP_016859236.1 | |
AK095284.1 | 1 | 53 | BAC04522.1 | |
AK296965.1 | 1 | 52 | ||
AK297262.1 | 1 | 47 | ||
AK302562.1 | ||||
AK311294.1 | ||||
AY358702.1 | 1 | 36 | AAQ89065.1 | |
BC146817.1 | 1 | 57 | ||
BP872689.1 | 1 | 67 | ||
BX647953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833648 | Chr.2:30283016 - 30473409 on Build GRCh38 | Gain | LOC285043 LCLAT1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |