Assay Details
Target Gene Details
Entrez Gene ID: | 83986 |
Gene Name: | family with sequence similarity 234 member A |
Gene Aliases: |
C16orf9, ITFG3, gs19 |
Location: |
Chr.16:234546-269943 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM234A | NR_104317.1 | |||
AK057165.1 | BAB71374.1 | |||
AK294581.1 |
Target Gene Details
Entrez Gene ID: | 8786 |
Gene Name: | regulator of G-protein signaling 11 |
Gene Aliases: |
RS11 |
Location: |
Chr.16:268301-275943 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS11 | NM_001286485.1 | 16 | 2310 | NP_001273414.1 |
NM_001286486.1 | 15 | 2178 | NP_001273415.1 | |
NM_003834.2 | 17 | 2354 | NP_003825.1 | |
NM_183337.2 | 17 | 2343 | NP_899180.1 | |
XM_011522719.2 | 18 | 2437 | XP_011521021.1 | |
XM_011522720.2 | 18 | 2377 | XP_011521022.1 | |
AB209582.1 | 9 | 4251 | BAD92819.1 | |
AF035153.1 | 17 | 2314 | AAC69175.1 | |
AF035154.1 | 17 | 2325 | AAC69176.1 | |
AK091701.1 | 15 | 2175 | ||
BC141803.1 | 17 | 2318 | ||
CK299324.1 | 1 | 80 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1479e59 | Chr.16:267152 - 268350 on Build GRCh38 | Duplication |
![]() ![]() |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3570031 | Chr.16:243389 - 278107 on Build GRCh38 | Gain |
![]() ![]() |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3637561 | Chr.16:261769 - 271795 on Build GRCh38 | Loss |
![]() ![]() |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs76735572] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)