Assay Details
Target Gene Details
Entrez Gene ID: | 7009 |
Gene Name: | transmembrane BAX inhibitor motif containing 6 |
Gene Aliases: |
BAXI1, BI-1, TEGT |
Location: |
Chr.12:49741510-49764934 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMBIM6 | NM_001098576.1 | NP_001092046.1 | ||
NM_003217.2 | NP_003208.2 | |||
XM_005269126.3 | XP_005269183.1 | |||
AF033095.1 | AAB87479.1 | |||
AK124849.1 | ||||
AK130050.1 | ||||
AK130462.1 | ||||
AK225804.1 | ||||
AK299587.1 | ||||
AK304577.1 | ||||
AK312238.1 | ||||
AY736129.1 | AAU29521.1 | |||
BC000916.1 | AAH00916.1 | |||
BC036203.1 | ||||
DA476870.1 | ||||
DC319835.1 | ||||
DC349385.1 | ||||
X75861.1 | CAA53472.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |