Assay Details
Target Gene Details
Entrez Gene ID: | 8405 |
Gene Name: | speckle type BTB/POZ protein |
Gene Aliases: |
BTBD32, TEF2 |
Location: |
Chr.17:49598884-49678163 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPOP | NM_001007226.1 | 12 | 2272 | NP_001007227.1 |
NM_001007227.1 | 11 | 2150 | NP_001007228.1 | |
NM_001007228.1 | 10 | 2098 | NP_001007229.1 | |
NM_001007229.1 | 11 | 2149 | NP_001007230.1 | |
NM_001007230.1 | 12 | 2201 | NP_001007231.1 | |
NM_003563.3 | 11 | 2220 | NP_003554.1 | |
XM_005257723.4 | 11 | 2444 | XP_005257780.1 | |
XM_005257724.4 | 10 | 2374 | XP_005257781.1 | |
XM_017025204.1 | 9 | 1887 | XP_016880693.1 | |
AK123385.1 | 1 | 1930 | ||
BQ226031.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |