Assay Details
Target Gene Details
Entrez Gene ID: | 64699 |
Gene Name: | transmembrane protease, serine 3 |
Gene Aliases: |
DFNB10, DFNB8, ECHOS1, TADG12 |
Location: |
Chr.21:42371887-42396846 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS3 | NM_001256317.1 | 13 | 1626 | NP_001243246.1 |
NM_024022.2 | 13 | 1629 | NP_076927.1 | |
NM_032404.2 | 10 | 1327 | NP_115780.1 | |
NR_046020.1 | 13 | 2384 | ||
AB038157.1 | 13 | 1629 | BAB20077.1 | |
AB038158.1 | 11 | 1715 | BAB20078.1 | |
AB038159.1 | 10 | 1296 | BAB20079.1 | |
AF201380.1 | 13 | 1574 | AAG37012.1 | |
AK131212.1 | 5 | 2160 | ||
AK172842.1 | 13 | 1589 | BAD18806.1 | |
AY358458.1 | 13 | 1550 | AAQ88823.1 | |
AY633572.1 | 13 | 2384 | AAT66641.1 | |
BC074846.2 | AAH74846.1 | |||
BC074847.2 | AAH74847.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |