Assay Details
Target Gene Details
Entrez Gene ID: | 10396 |
Gene Name: | ATPase phospholipid transporting 8A1 |
Gene Aliases: |
ATPASEII, ATPIA, ATPP2 |
Location: |
Chr.4:42408373-42657134 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 37 - Exon 38 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP8A1 | NM_001105529.1 | NP_001098999.1 | ||
NM_006095.2 | NP_006086.1 | |||
XM_005248043.2 | XP_005248100.1 | |||
XM_011513615.1 | XP_011511917.1 | |||
XM_011513616.2 | XP_011511918.1 | |||
XM_011513618.1 | XP_011511920.1 | |||
XM_017007645.1 | XP_016863134.1 | |||
XM_017007646.1 | XP_016863135.1 | |||
XM_017007647.1 | XP_016863136.1 | |||
AB013452.1 | BAA77248.1 | |||
AB209687.1 | BAD92924.1 | |||
AF067820.1 | AAD34706.1 | |||
BC109317.1 | AAI09318.1 | |||
BC109318.1 | AAI09319.1 | |||
BX648675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv998958 | Chr.4:42404571 - 42525872 on Build GRCh38 | Gain | ATP8A1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |