Assay Details
Target Gene Details
Entrez Gene ID: | 4793 |
Gene Name: | NFKB inhibitor beta |
Gene Aliases: |
IKBB, TRIP9 |
Location: |
Chr.19:38899503-38908894 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFKBIB | NM_001243116.1 | NP_001230045.1 | ||
NM_002503.4 | NP_002494.2 | |||
NR_040515.1 | 5 | 2032 | ||
XM_006723226.2 | XP_006723289.2 | |||
XM_006723227.3 | XP_006723290.3 | |||
AA044256.1 | 1 | 152 | ||
AI684894.1 | ||||
AK057862.1 | 5 | 1983 | BAB71599.1 | |
BC015528.1 | AAH15528.1 | |||
BI520169.1 | ||||
BT006743.1 | AAP35389.1 | |||
BU634404.1 | ||||
BX111397.1 | 1 | 158 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |