Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_177533.4 | 5 | 750 | NP_803877.2 |
AB087802.1 | 5 | 636 | BAC65455.1 | |
AK128196.1 | 7 | 3215 | ||
BC041584.1 | 5 | 712 | AAH41584.1 | |
BE671270.1 | 1 | 122 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | JAG2 LOC102723354 LOC100507437 PACS2 CRIP2 MIR6765 MTA1 BRF1 GPR132 BTBD6 TEX22 NUDT14 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | LOC105370697 JAG2 IGHA2 C14orf80 PACS2 CRIP1 CRIP2 MTA1 BTBD6 NUDT14 LOC100507437 TMEM121 MIR6765 IGH BRF1 TEX22 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | JAG2 LOC102723354 MIR6765 BRF1 GPR132 NUDT14 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | JAG2 MIR6765 BRF1 NUDT14 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | CKB MARK3 LOC105370691 C14orf79 INF2 C14orf180 TDRD9 BAG5 APOPT1 ZFYVE21 SIVA1 CEP170B KIF26A LOC107984648 LOC105378183 LINC00637 ASPG CDCA4 BRF1 LOC105370708 XRCC3 LOC388022 C14orf2 RD3L JAG2 MIR203B SNORA28 LOC101929634 AHNAK2 PACS2 TMEM179 ZBTB42 GPR132 ADSSL1 BTBD6 NUDT14 MIR4710 PPP1R13B LOC102723354 TRMT61A PLD4 MIR203A LINC00638 MIR6765 EIF5 KLC1 AKT1 LOC105370687 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | JAG2 LOC102723354 AHNAK2 C14orf79 MIR6765 CDCA4 BRF1 GPR132 BTBD6 NUDT14 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGHV4-55 IGHV1-69 MIR4538 IGHA2 CRIP2 IGHV3-41 MTA1 ELK2AP IGHG4 LINC00221 IGHV3-23 IGHV3-30 IGHV3-71 KIAA0125 IGHV3-69-1 MIR4539 IGHG3 IGHV3-48 IGHV4-31 IGHD MIR4537 IGH BRF1 MIR8071-1 IGHM TEX22 LOC105370697 JAG2 MIR8071-2 C14orf80 PACS2 LINC00226 CRIP1 IGHG2 ADAM6 BTBD6 NUDT14 LOC100507437 IGHG1 TMEM121 MIR4507 LOC107987211 MIR6765 IGHA1 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | JAG2 LOC102723354 AHNAK2 C14orf79 MIR6765 CDCA4 BRF1 GPR132 NUDT14 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |