Assay Details
Target Gene Details
Entrez Gene ID: | 9228 |
Gene Name: | DLG associated protein 2 |
Gene Aliases: |
DAP2, SAPAP2 |
Location: |
Chr.8:1501366-1708476 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP2 | NM_001277161.1 | NP_001264090.1 | ||
NM_004745.4 | NP_004736.2 | |||
AF009204.1 | AAB71661.1 | |||
AF119817.1 | AAF34344.1 | |||
AF119818.1 | AAF34345.1 | |||
BC141862.1 | ||||
KJ534823.1 |
Target Gene Details
Entrez Gene ID: | 100507435 |
Gene Name: | DLGAP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:1565509-1621664 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP2-AS1 | NR_103863.1 | |||
AW303348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950273 | Chr.8:1561235 - 1573734 on Build GRCh38 | Deletion | DLGAP2 DLGAP2-AS1 |
nsv6045 | Chr.8:1542683 - 1577255 on Build GRCh38 | Insertion | DLGAP2 DLGAP2-AS1 |
nsv469881 | Chr.8:1426471 - 1613783 on Build GRCh38 | Loss | DLGAP2 DLGAP2-AS1 |
nsv1024325 | Chr.8:651043 - 3203837 on Build GRCh38 | Gain | DLGAP2 LOC105377776 LOC101927815 LOC100131395 KBTBD11-OT1 MIR3674 LOC286083 LOC101928058 LOC101927752 ERICH1 LOC401442 MIR596 CLN8 MIR7160 MYOM2 LOC105377792 LOC105377777 ARHGEF10 ERICH1-AS1 KBTBD11 DLGAP2-AS1 LOC105377774 LOC105377781 CSMD1 |
esv3576504 | Chr.8:951237 - 1906751 on Build GRCh38 | Gain | DLGAP2 LOC105377776 ARHGEF10 ERICH1-AS1 MIR3674 DLGAP2-AS1 LOC286083 LOC101927752 MIR596 CLN8 |
nsv482921 | Chr.8:1545246 - 1709424 on Build GRCh38 | Gain | DLGAP2 DLGAP2-AS1 |
nsv609509 | Chr.8:1352944 - 1641722 on Build GRCh38 | Gain | DLGAP2 LOC105377776 DLGAP2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |