Assay Details
Target Gene Details
Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902799 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH4A1 | NM_001161504.1 | NP_001154976.1 | ||
NM_001319218.1 | NP_001306147.1 | |||
NM_003748.3 | NP_003739.2 | |||
NM_170726.2 | NP_733844.1 | |||
AK222486.1 | BAD96206.1 | |||
AK289972.1 | ||||
AK294552.1 | ||||
BC007581.1 | AAH07581.1 | |||
BC023600.2 | AAH23600.1 | |||
BX649081.1 | 1 | 875 | ||
FJ462711.1 | ||||
U24266.1 | AAC50500.1 | |||
U24267.1 | AAC50501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545719 | Chr.1:18873686 - 18881699 on Build GRCh38 | Loss | ALDH4A1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |