Assay Details
Target Gene Details
Entrez Gene ID: | 375607 |
Gene Name: | N-acetyltransferase 16 (putative) |
Gene Aliases: |
C7orf52 |
Location: |
Chr.7:101170493-101180276 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAT16 | NM_198571.2 | 4 | 1293 | NP_940973.2 |
XM_005250325.1 | 5 | 1272 | XP_005250382.1 | |
XM_011516168.1 | 4 | 1254 | XP_011514470.1 | |
AK092119.1 | ||||
AK096556.1 | 4 | 1293 | BAC04815.1 | |
AL831911.1 | 2 | 563 | CAD38574.3 | |
BC104841.1 | 5 | 1272 | AAI04842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422376 | Chr.7:101100204 - 101373190 on Build GRCh38 | Duplication | LOC101927721 ZNHIT1 NAT16 MOGAT3 PLOD3 MIR4653 FIS1 COL26A1 CLDN15 AP1S1 LOC101927746 IFT22 VGF SERPINE1 |
nsv1030604 | Chr.7:101074451 - 101580185 on Build GRCh38 | Gain | LOC101927721 ZNHIT1 NAT16 LINC01007 MOGAT3 PLOD3 MIR4653 FIS1 COL26A1 CLDN15 AP1S1 LOC101927746 IFT22 VGF SERPINE1 TRIM56 |
dgv11534n54 | Chr.7:101160859 - 101238359 on Build GRCh38 | Loss | PLOD3 ZNHIT1 CLDN15 AP1S1 VGF NAT16 MOGAT3 |
esv2752134 | Chr.7:101156850 - 101498586 on Build GRCh38 | Gain | LOC101927721 ZNHIT1 NAT16 MOGAT3 PLOD3 MIR4653 FIS1 COL26A1 CLDN15 AP1S1 LOC101927746 IFT22 VGF |
nsv607973 | Chr.7:101139179 - 101230554 on Build GRCh38 | Loss | PLOD3 MIR4653 ZNHIT1 AP1S1 VGF SERPINE1 NAT16 MOGAT3 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | AGFG2 ZNHIT1 ACHE POP7 PVRIG2P AZGP1 GAL3ST4 PVRIG FIS1 AP1S1 SERPINE1 STAG3L5P-PVRIG2P-PILRB ZCWPW1 MIR93 FBXO24 LOC102724094 MEPCE LOC105375429 MIR6875 PCOLCE-AS1 ZASP SAP25 AZGP1P1 ZNF3 SLC12A9 PCOLCE MUC17 MOGAT3 C7orf61 ZAN GPC2 MIR4653 VGF MUC3A MIR4658 ACTL6B PMS2P1 SPDYE3 GIGYF1 UFSP1 TRIP6 PILRA TAF6 ZKSCAN1 EPO STAG3L5P GNB2 PILRB NAT16 EPHB4 MOSPD3 AP4M1 PLOD3 ZSCAN21 COPS6 MBLAC1 LRCH4 MIR6840 NYAP1 PPP1R35 STAG3 SRRT MCM7 C7orf43 GATS TSC22D4 TFR2 MIR25 MIR106B LAMTOR4 MUC12 CLDN15 CNPY4 TRIM56 |
nsv831075 | Chr.7:101049016 - 101273737 on Build GRCh38 | Loss | PLOD3 MIR4653 FIS1 ZNHIT1 CLDN15 AP1S1 VGF SERPINE1 TRIM56 NAT16 MUC17 MOGAT3 |
nsv607974 | Chr.7:101149529 - 101328323 on Build GRCh38 | Loss | LOC101927721 PLOD3 MIR4653 FIS1 ZNHIT1 CLDN15 AP1S1 LOC101927746 IFT22 VGF NAT16 MOGAT3 |
nsv1018841 | Chr.7:101117129 - 101225018 on Build GRCh38 | Gain | PLOD3 MIR4653 ZNHIT1 AP1S1 VGF SERPINE1 NAT16 MOGAT3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |