Assay Details
Target Gene Details
Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902799 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH4A1 | NM_001161504.1 | 15 | 2203 | NP_001154976.1 |
NM_001319218.1 | 14 | 2190 | NP_001306147.1 | |
NM_003748.3 | 15 | 2343 | NP_003739.2 | |
NM_170726.2 | NP_733844.1 | |||
AK222486.1 | 15 | 2099 | BAD96206.1 | |
BC007581.1 | 15 | 2073 | AAH07581.1 | |
BC023600.2 | 15 | 2084 | AAH23600.1 | |
BX649081.1 | 2 | 1448 | ||
FJ462711.1 | 14 | 1928 | ||
U24266.1 | 15 | 2097 | AAC50500.1 | |
U24267.1 | AAC50501.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |