Assay Details
Target Gene Details
Entrez Gene ID: | 83733 |
Gene Name: | solute carrier family 25 member 18 |
Gene Aliases: |
GC2 |
Location: |
Chr.22:17562470-17590890 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | LOC101929372 CECR2 BCL2L13 SLC25A18 ATP6V1E1 |
nsv1065055 | Chr.22:17559085 - 17632626 on Build GRCh38 | Gain | LOC101929372 BCL2L13 SLC25A18 ATP6V1E1 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | LOC101929372 LINC00528 BID CECR2 BCL2L13 MICAL3 MIR3198-1 SLC25A18 ATP6V1E1 |
nsv3553 | Chr.22:17553601 - 17608096 on Build GRCh38 | Deletion | LOC101929372 CECR2 SLC25A18 ATP6V1E1 |
nsv510802 | Chr.22:17550100 - 17604891 on Build GRCh38 | Deletion | LOC101929372 CECR2 SLC25A18 ATP6V1E1 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC101929372 PI4KAP1 TUBA8 SLC25A18 ATP6V1E1 LINC00528 BID FLJ41941 BCL2L13 MICAL3 LOC100996432 LOC642643 PEX26 MIR648 LOC729461 TMEM191B FAM230A LOC102725072 RIMBP3 CECR2 LOC100996415 LOC100996401 USP18 GGT3P MIR3198-1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |