Assay Details
Target Gene Details
Entrez Gene ID: | 51738 |
Gene Name: | ghrelin/obestatin prepropeptide |
Gene Aliases: |
MTLRP |
Location: |
Chr.3:10285750-10292947 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Target Gene Details
Entrez Gene ID: | 100126793 |
Gene Name: | ghrelin opposite strand/antisense RNA |
Gene Aliases: |
GHRL-AS1, GHRLAS, NCRNA00068 |
Location: |
Chr.3:10280952-10293449 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRLOS | NR_004431.3 | |||
NR_024144.2 | ||||
NR_024145.2 | ||||
NR_073566.1 | ||||
NR_073567.1 | ||||
NR_073568.1 | ||||
EF549568.1 | ||||
EF549570.1 | ||||
EU072081.1 | ||||
EU072082.1 | ||||
EU789531.1 | ||||
EU789534.1 | ||||
EU789535.1 | ||||
EU789536.1 | ||||
EU789537.1 | ||||
EU789538.1 | ||||
EU789539.1 | ||||
EU789540.1 | ||||
EU789541.1 | ||||
EU789542.1 | ||||
EU789543.1 | ||||
EU789544.1 | ||||
EU789545.1 | ||||
EU789546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | VGLL4 VHL MIR378B GHRLOS ATP2B2-IT2 LINC00606 SLC6A1-AS1 SLC6A11 ATP2B2 FANCD2 MIR885 BRK1 LOC101927467 LINC00852 SEC13 TATDN2 HRH1 FANCD2OS SLC6A1 GHRL ATG7 IRAK2 TAMM41 |
nsv508903 | Chr.3:10275353 - 10401894 on Build GRCh38 | Insertion | ATP2B2 MIR885 MIR378B GHRLOS LINC00852 SEC13 TATDN2 GHRL |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | GHRLOS LINC00852 TATDN2 GHRL |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | VGLL4 VHL GHRLOS LINC00606 ATP2B2 MIR885 EMC3 BRK1 LOC101927467 SEC13 FANCD2OS SLC6A1 GHRL MIR378B CIDECP ATP2B2-IT2 SLC6A1-AS1 SLC6A11 LOC401052 FANCD2 FANCD2P2 LINC00852 TATDN2 HRH1 ATG7 IRAK2 TAMM41 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | VGLL4 VHL MIR378B GHRLOS ATP2B2-IT2 LINC00606 SLC6A1-AS1 SLC6A11 ATP2B2 FANCD2 MIR885 FANCD2P2 BRK1 LOC101927467 LINC00852 SEC13 TATDN2 HRH1 FANCD2OS SLC6A1 GHRL ATG7 IRAK2 TAMM41 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | VGLL4 VHL MIR378B GHRLOS ATP2B2-IT2 LINC00606 SLC6A1-AS1 SLC6A11 ATP2B2 FANCD2 MIR885 FANCD2P2 BRK1 LOC101927467 LINC00852 SEC13 TATDN2 HRH1 FANCD2OS SLC6A1 GHRL ATG7 IRAK2 TAMM41 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | VGLL4 EMC3-AS1 VHL GHRLOS LINC00606 ATP2B2 MIR885 EMC3 BRK1 LOC101927467 SEC13 FANCD2OS SLC6A1 GHRL MIR378B CIDECP ATP2B2-IT2 SLC6A1-AS1 SLC6A11 LOC401052 FANCD2 FANCD2P2 LINC00852 TATDN2 HRH1 ATG7 IRAK2 TAMM41 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |