Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | 5 | 1035 | NP_848611.2 |
AK092858.1 | 5 | 927 | BAC03991.1 | |
AK092906.1 | 5 | 799 | BAC04000.1 | |
AK292004.1 | 5 | 944 | ||
AK300915.1 | 5 | 820 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | DUS2 FBXL8 SLC9A5 CTRL TRADD E2F4 TMEM208 THAP11 DDX28 LRRC29 KIAA0895L DPEP3 LRRC36 ATP6V0D1 SLC12A4 LOC100505942 NRN1L PSKH1 AGRP PLEKHG4 CTCF LCAT MIR328 FAM65A TSNAXIP1 NOL3 LOC105369155 CENPT PSMB10 HSD11B2 ELMO3 LOC100131303 EXOC3L1 HSF4 C16orf86 DPEP2 GFOD2 TPPP3 EDC4 RANBP10 KCTD19 LOC107984813 FHOD1 PARD6A ACD ZDHHC1 CARMIL2 ENKD1 NUTF2 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | FHOD1 FBXL8 ELMO3 PLEKHG4 SLC9A5 EXOC3L1 HSF4 TRADD C16orf70 E2F4 TMEM208 MIR328 NOL3 LOC105369155 LRRC29 KIAA0895L B3GNT9 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | FHOD1 FBXL8 ELMO3 EXOC3L1 HSF4 TRADD C16orf70 E2F4 TMEM208 MIR328 NOL3 LOC105369155 LRRC29 KIAA0895L B3GNT9 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | E2F4 MIR328 FBXL8 NOL3 ELMO3 LOC105369155 LRRC29 KIAA0895L EXOC3L1 HSF4 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | E2F4 TMEM208 MIR328 FHOD1 ELMO3 LOC105369155 LRRC29 PLEKHG4 KIAA0895L SLC9A5 EXOC3L1 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | KCTD19 FHOD1 ELMO3 PLEKHG4 SLC9A5 EXOC3L1 E2F4 TMEM208 MIR328 NOL3 LOC105369155 LRRC29 KIAA0895L |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | DUS2 FBXL8 SLC9A5 CTRL TRADD C16orf70 E2F4 TMEM208 THAP11 DDX28 LRRC29 KIAA0895L DPEP3 LRRC36 ATP6V0D1 SLC12A4 LOC100505942 NRN1L PSKH1 AGRP PLEKHG4 CTCF LCAT MIR328 FAM65A TSNAXIP1 NOL3 LOC105369155 CENPT PSMB10 HSD11B2 ELMO3 LOC100131303 EXOC3L1 HSF4 C16orf86 DPEP2 GFOD2 TPPP3 EDC4 B3GNT9 RANBP10 KCTD19 LOC107984813 FHOD1 PARD6A ACD ZDHHC1 CARMIL2 ENKD1 NUTF2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |