Assay Details
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | 1 | 1640 | ||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 | |||
DB041321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 LOC107984648 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | XRCC3 ZFYVE21 LOC107984648 KLC1 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | XRCC3 ZFYVE21 LOC107984648 KLC1 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | XRCC3 ZFYVE21 LOC107984648 KLC1 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LINC00638 LOC101929634 AHNAK2 LOC105370708 LOC102723354 EIF5 ASPG KIF26A ZFYVE21 C14orf2 LOC388022 LOC105370691 CEP170B PPP1R13B MIR4710 APOPT1 MARK3 PLD4 C14orf79 BAG5 JAG2 XRCC3 PACS2 LOC105370687 NUDT14 CKB SNORA28 MIR6765 SIVA1 C14orf180 BRF1 LOC107984648 BTBD6 LINC00637 KLC1 ZBTB42 ADSSL1 INF2 RD3L GPR132 TDRD9 CDCA4 AKT1 MIR203B TRMT61A TMEM179 LOC105378183 MIR203A |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | RD3L TDRD9 XRCC3 ZFYVE21 C14orf2 LOC107984648 PPP1R13B LINC00637 KLC1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |