Assay Details
Target Gene Details
Entrez Gene ID: | 5521 |
Gene Name: | protein phosphatase 2 regulatory subunit Bbeta |
Gene Aliases: |
B55BETA, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55-BETA, PR55BETA, SCA12 |
Location: |
Chr.5:146589504-147081520 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP2R2B | NM_001271899.1 | 10 | 1886 | NP_001258828.1 |
NM_001271900.1 | 11 | 2080 | NP_001258829.1 | |
NM_001271948.1 | 10 | 2413 | NP_001258877.1 | |
NM_181674.2 | 10 | 2211 | NP_858060.2 | |
NM_181675.3 | 10 | 1933 | NP_858061.2 | |
NM_181676.2 | 9 | 2017 | NP_858062.1 | |
NM_181677.2 | 11 | 1843 | NP_858063.1 | |
NM_181678.2 | 11 | 1856 | NP_858064.1 | |
NR_073526.1 | 9 | 2328 | ||
NR_073527.1 | 10 | 1965 | ||
AI621232.1 | 1 | 166 | ||
AK056192.1 | 10 | 1893 | ||
AK294659.1 | ||||
AK295347.1 | 10 | 1857 | ||
BC031790.1 | 10 | 2101 | AAH31790.1 | |
BX647887.1 | 10 | 1947 | ||
M64930.1 | 9 | 2139 | AAA36491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5052 | Chr.5:146559163 - 146592780 on Build GRCh38 | Insertion | PPP2R2B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |