Assay Details
Target Gene Details
Entrez Gene ID: | 6440 |
Gene Name: | surfactant protein C |
Gene Aliases: |
BRICD6, PSP-C, SFTP2, SMDP2, SP-C |
Location: |
Chr.8:22161671-22164479 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SFTPC | NM_001172357.1 | 2 | 322 | NP_001165828.1 |
NM_001172410.1 | 2 | 322 | NP_001165881.1 | |
NM_001317778.1 | 2 | 322 | NP_001304707.1 | |
NM_001317779.1 | NP_001304708.1 | |||
NM_001317780.1 | 2 | 322 | NP_001304709.1 | |
NM_003018.3 | 2 | 322 | NP_003009.2 | |
XM_011544613.2 | XP_011542915.1 | |||
AK058094.1 | 5 | 1014 | ||
AK298119.1 | 2 | 193 | ||
AK315742.1 | 2 | 196 | ||
BC005913.1 | 2 | 195 | AAH05913.1 | |
BG535994.1 | 2 | 199 | ||
BG569816.1 | 2 | 199 | ||
BI490051.1 | ||||
BI524154.1 | 2 | 179 | ||
BM993053.1 | ||||
DQ884411.1 | 2 | 164 | ||
J03517.1 | 2 | 191 | AAA36634.1 | |
J03553.1 | 2 | 338 | AAA36631.1 | |
KU178329.1 | 2 | 164 | ||
KU178330.1 | 2 | 164 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv465613 | Chr.8:22046701 - 22197835 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv528590 | Chr.8:22034428 - 22168131 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv527293 | Chr.8:22106867 - 22163524 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv831263 | Chr.8:22031088 - 22184413 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470204 | Chr.8:22094715 - 22171155 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv12054n54 | Chr.8:22045805 - 22251098 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv831264 | Chr.8:22093700 - 22328040 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv610771 | Chr.8:22149780 - 22200609 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)