Assay Details
Target Gene Details
Entrez Gene ID: | 2259 |
Gene Name: | fibroblast growth factor 14 |
Gene Aliases: |
FGF-14, FHF-4, FHF4, SCA27 |
Location: |
Chr.13:101720855-102402428 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF14 | NM_001321931.1 | 6 | 1138 | NP_001308860.1 |
NM_001321932.1 | 6 | 1224 | NP_001308861.1 | |
NM_001321933.1 | 6 | 1134 | NP_001308862.1 | |
NM_001321934.1 | 5 | 897 | NP_001308863.1 | |
NM_001321935.1 | 5 | 750 | NP_001308864.1 | |
NM_001321936.1 | 6 | 1077 | NP_001308865.1 | |
NM_001321938.1 | 7 | 1125 | NP_001308867.1 | |
NM_001321939.1 | 4 | 1132 | NP_001308868.1 | |
NM_001321940.1 | 6 | 1480 | NP_001308869.1 | |
NM_001321941.1 | 6 | 1312 | NP_001308870.1 | |
NM_001321942.1 | 6 | 1319 | NP_001308871.1 | |
NM_001321943.1 | 6 | 1356 | NP_001308872.1 | |
NM_001321944.1 | 6 | 1572 | NP_001308873.1 | |
NM_001321945.1 | 6 | 1115 | NP_001308874.1 | |
NM_001321946.1 | 7 | 1221 | NP_001308875.1 | |
NM_001321947.1 | 6 | 1039 | NP_001308876.1 | |
NM_001321948.1 | 6 | 1111 | NP_001308877.1 | |
NM_001321949.1 | 6 | 1808 | NP_001308878.1 | |
NM_004115.3 | 5 | 874 | NP_004106.1 | |
NM_175929.2 | 5 | 985 | NP_787125.1 | |
AY188178.1 | AAO31806.1 | |||
BC100920.1 | AAI00921.1 | |||
BC100921.1 | AAI00922.1 | |||
BC100922.2 | AAI00923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832702 | Chr.13:101621711 - 101795212 on Build GRCh38 | Gain | FGF14 ITGBL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |