Abnova
Manufacturer Code:H00010781B01
Catalog # 89-007-054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZNF266 (AAH17407.1 1 a.a. ~ 263 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ZNF266 protein.
For Research Use Only
Protein Aliases: HZF1; zinc finger protein 1; Zinc finger protein 266; Zinc finger protein HZF1
Gene Aliases: HZF1; KIAA2007; ZNF266
UniProt ID: (Human) Q14584
Entrez Gene ID: (Human) 10781
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.