Abnova
Manufacturer Code:H00080345B01
Catalog # 89-008-249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZNF435 (NP_079507.1 1 a.a. ~ 348 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ZNF435 protein.
For Research Use Only
Protein Aliases: dJ265C24.3; FLJ22191; Zinc finger and SCAN domain-containing protein 16; Zinc finger protein 392; Zinc finger protein 435; ZNF392; ZNF435
Gene Aliases: dJ265C24.3; ZNF392; ZNF435; ZSCAN16
UniProt ID: (Human) Q9H4T2
Entrez Gene ID: (Human) 80345
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.