Novus Biologicals
Manufacturer Code:NB300317B
Catalog # NB300317B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This xCT Antibody was prepared from a synthetic peptide made to a region within the N-terminus of the mouse xCT protein (between residues 1-50). [Uniprot: Q9WTR6] |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amino acid transport system xc-; Amino acid transport system xc- Calcium channel blocker resistance protein CCBR1 cystine/glutamate transporter Member 11 (Slc7a11) Solute carrier family 7 member 11 solute carrier family 7 (cationic amino acid transporter y+ system) member 11 xCTCCBR1; Calcium channel blocker resistance protein CCBR1; Cystine/glutamate transporter; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; Solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; xCT
Gene Aliases: CCBR1; SLC7A11; xCT
UniProt ID: (Human) Q9UPY5
Entrez Gene ID: (Human) 23657
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.