Abnova
Manufacturer Code:H00023063B01
Catalog # 89-007-234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIAA0261 (ENSP00000263070 1 a.a. ~ 402 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human KIAA0261 protein.
For Research Use Only
Protein Aliases: FOE; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; KIAA0261; WAPL; WAPL cohesin release factor; Wings apart-like protein homolog
Gene Aliases: FOE; KIAA0261; WAPAL; WAPL
UniProt ID: (Human) Q7Z5K2
Entrez Gene ID: (Human) 23063
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.