Proteintech
Manufacturer Code:169221AP150UL
Catalog # 50-173-1261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | vitamin D binding protein Fusion Protein Ag10286 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBP; DBP DBP/GC GC Gc globulin Group specific component VDB VDBG VDBP Vitamin D binding protein; DBP-maf; epididymis secretory protein Li 51; Gc; Gc protein-derived macrophage activating factor; Gc-globulin; Gc-MAF; Group-specific component; group-specific component (vitamin D binding protein); VDB; vitamin D-binding alpha-globulin; Vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor
Gene Aliases: DBP; DBP/GC; DBP02; GC; Gc-MAF; GcMAF; GRD3; HEL-S-51; VDB; VDBG; VDBP
UniProt ID: (Human) P02774, (Mouse) P21614
Entrez Gene ID: (Human) 2638, (Mouse) 14473, (Rat) 24384
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.