Invitrogen
Manufacturer Code:PA575222
Catalog # PIPA575222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 221-270 of Human v-Myb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-Myb; Myb-like protein 1; Myb-related protein A; Myb-related protein A A-Myb Myb-like protein 1 MYBL1 AMYB; v-myb avian myeloblastosis viral oncogene homolog-like 1
Gene Aliases: A-MYB; AMYB; G1-419-6; MYBL1; repro9
UniProt ID: (Human) P10243, (Mouse) P51960
Entrez Gene ID: (Human) 4603, (Rat) 297783, (Mouse) 17864
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.