Abnova
Manufacturer Code:H00054963B01
Catalog # 89-007-779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | UCKL1 (AAH33078.1 1 a.a. ~ 419 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human UCKL1 protein.
For Research Use Only
Protein Aliases: UCK1-LIKE; UCK1L; uridine-cytidine kinase 1-like 1; Uridine-cytidine kinase-like 1; URKL1
Gene Aliases: F538; UCK1L; UCKL1; URKL1
UniProt ID: (Human) Q9NWZ5
Entrez Gene ID: (Human) 54963
Molecular Function:
glycosyltransferase
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.