Invitrogen
Manufacturer Code:PA579826
Catalog # PIPA579826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat uPA Receptor recombit protein (Position: L25-R261). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD87; plasminogen activator urokinase receptor 3; U-PAR; urinary plasminogen activator receptor 2; urinary plasminogen activator receptor 3; urokinase plasminogen activator receptor; Urokinase plasminogen activator surface receptor; Urokinase plasminogen activator surface receptor U-PAR uPAR CD87 Plaur; urokinase-type plasminogen activator receptor
Gene Aliases: Cd87; Par; Plaur; Plaur3; u-PAR; uPAR; uPAR-2; uPAR-3
UniProt ID: (Rat) P49616, (Mouse) P35456
Entrez Gene ID: (Rat) 50692, (Mouse) 18793
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.