Invitrogen
Manufacturer Code:PA581629
Catalog # PIPA581629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Rat PLAUR/CD87 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD87; plasminogen activator urokinase receptor 3; PLAUR U-PAR UPAR URKR monocyte activation antigen Mo3 u-plasminogen activator receptor form 2 urokinase plasminogen activator surface receptor; U-PAR; urinary plasminogen activator receptor 2; urinary plasminogen activator receptor 3; urokinase plasminogen activator receptor; Urokinase plasminogen activator surface receptor
Gene Aliases: Par; Plaur; Plaur3; uPAR; uPAR-2; uPAR-3
UniProt ID: (Rat) P49616
Entrez Gene ID: (Rat) 50692
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.