Novus Biologicals
Manufacturer Code:NBP214711
Catalog # NBP214711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: VRLWEEGEELELVEKSCTHSEKTNRTLSYRTGLKITSLTEVVCGLDLCNQ GNSGRAVTYSRSRYLECISCGSSDMSCERGRHQSLQCRSPEEQCLDVVTH WIQEGE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD87; CD87 CD87 antigen Monocyte activation antigen Mo3 plasminogen activator urokinase receptor uPAR U-PAR UPARurokinase plasminogen activator surface receptor u-plasminogen activator receptor form 2 URKRMO3; Monocyte activation antigen Mo3; U-PAR; u-plasminogen activator receptor form 2; Urokinase plasminogen activator surface receptor; urokinase-type plasminogen activator (uPA) receptor
Gene Aliases: CD87; MO3; PLAUR; U-PAR; UPAR; URKR
UniProt ID: (Human) Q03405
Entrez Gene ID: (Human) 5329
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.