R&D Systems
Manufacturer Code:AF534
Catalog # AF534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1 Leu24-Thr297 Accession # Q545X5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD87; CD87 CD87 antigen Monocyte activation antigen Mo3 plasminogen activator urokinase receptor PLAUR uPAR U-PAR UPARurokinase plasminogen activator surface receptor u-plasminogen activator receptor form 2 URKRMO3; Monocyte activation antigen Mo3; U-PAR; u-plasminogen activator receptor form 2; Urokinase plasminogen activator surface receptor; urokinase-type plasminogen activator (uPA) receptor
Gene Aliases: CD87; MO3; PLAUR; U-PAR; UPAR; URKR
UniProt ID: (Human) Q03405
Entrez Gene ID: (Human) 5329
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.