Invitrogen
Manufacturer Code:PA138835
Catalog # 01-674-774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from C-terminus of mouse iNOS and nNOS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bNOS; cNOS; Constitutive NOS; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; HEP-NOS; HEP-NOS INOS NOS NOS2A; Hepatocyte NOS; Inducible NO synthase; Inducible NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxide synthase 1 (neuronal); nitric oxide synthase 2, inducible; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; Nitric oxide synthase, brain; Nitric oxide synthase, endothelial; Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; NOS type I; NOS type II; NOS type III; NOS, type II; NOSIII; Peptidyl-cysteine S-nitrosylase NOS1; Peptidyl-cysteine S-nitrosylase NOS2
Gene Aliases: 2310065A03Rik; bNOS; ECNOS; eNOS; HEP-NOS; IHPS1; INOS; N-NOS; NC-NOS; nNOS; NOS; Nos-3; NOS1; NOS2; NOS2A; NOS3
UniProt ID: (Human) P29475, (Human) P35228, (Human) P29474, (Mouse) P70313, (Rat) Q62600
Entrez Gene ID: (Human) 4842, (Human) 4843, (Human) 4846, (Mouse) 18127, (Rat) 24600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.