Novus Biologicals
Manufacturer Code:NBP223504AF750
Catalog # NP22304A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | U-6 |
Immunogen | Native human uPA |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ATF EC 3.4.21 EC 3.4.21.73 plasminogen activator urokinase uPA u-PA U-plasminogen activator urinary urokinase-type plasminogen activator; plasminogen activator, urinary; U-plasminogen activator; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A
Gene Aliases: ATF; BDPLT5; PLAU; QPD; u-PA; UPA; URK
UniProt ID: (Human) P00749
Entrez Gene ID: (Human) 5328
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.