Abnova
Manufacturer Code:H00007167M02A
Catalog # 89-001-491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C3 |
Immunogen | TPI1 (AAH07086.1 1 a.a. ~ 249 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant TPI1.
For Research Use Only
Protein Aliases: epididymis secretory protein Li 49; Methylglyoxal synthase; MGC88108; TIM; TPI; Triose-phosphate isomerase; Triosephosphate isomerase
Gene Aliases: HEL-S-49; TIM; TPI; TPI1; TPID
UniProt ID: (Human) P60174
Entrez Gene ID: (Human) 7167
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.